missing translation for 'onlineSavingsMsg'
Learn More

Melford Genistein

Product Code. 30018288
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018288 25 mg 25mg
30018289 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018288 Supplier Melford Supplier No. G360000.025
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibits protein tyrosine kinases.

Inhibits protein tyrosine kinases.

Specifications

CAS 446-72-0
Quantity 25 mg
Molecular Formula C15H10O5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.