missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Genistein

Product Code. 17110012
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17110012 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17110012 Supplier STEMCELL Technologies Supplier No. 72002
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

NF-kB, AMPK, and tyrosine kinase pathway inhibitor; Inhibits tyrosine kinase, topoisomerase II and activates PPARγ

  • Genistein is a phytoestrogen that exhibits a wide range of biological effects
  • It acts by competing for the ATP-binding site of tyrosine kinases but does not inhibit the activity of serine- and threonine-specific kinases such as cAMP-dependent protein kinase, protein kinase C, and phosphorylase kinase (Akiyama et al.) Genistein binds to and activates peroxisome proliferator-activated receptor-gamma (PPARγ) and estrogen receptors, leading to a decrease in osteogenesis and an increase in adipogenesis (Dang et al.) Genistein also binds and activates the seven-transmembrane estrogen receptor GPR30 to activate alternative estrogen signaling pathways (Thomas & Dong)
  • Inhibits adipocyte differentiation in 3T3-L1 cell culture (Hwang et al.)
  • Control of osteoblast and adipocyte cell fates in mouse bone marrow cells (Dang et al.)
  • Inhibits growth of leukemia, lymphoma, neuroblastoma, gastric, breast, and prostate cancer cells (Sarkar & Li)

Specifications

CAS 446-72-0
Quantity 100 mg
Molecular Formula C15H10O5
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.