missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scientific Industries Genie-Vortex 2 Replacement On/Off/On Switch
Click to view available options
For Use With (Equipment):
Vortex-Genie 2
Unit Size:
Each
Description
- Easily installed replacement On/Off/On toggle switch
Specifications
Specifications
| Product Type | Replacement On/Off/On Switch |
| Color | Silver |
| For Use With (Equipment) | Vortex-Genie 2 |
| Packaging Quantity | 1 Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction