missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ Installation Standard DS-33 , [6FAM™, VIC™, NED™, PET™ dyes]

Product Code. 15859766 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859766 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15859766 Supplier Applied Biosystems™ Supplier No. 4330397
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

4 Tubes of fluorescently labeled pooled PCR products, 1 tube of GeneScan -500 LIZ Size Standard, and 1 tube of loading buffer

Using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in CEPH individual 1347.

The DS-33 GeneScan™ Installation Standards using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in Human DNA. The kit also contains a tube of GeneScan™ 500 LIZ™ Size Standard.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains 4 tubes of fluorescently labeled pooled PCR products and 1 tube of GeneScan 500 LIZ Size Standard.

Store the kit at -15 to -25°C until ready to use. Upon thawing, store the GeneScan 500 LIZ Size Standard tube at 2-8°C.

For Use With (Equipment) 3730 DNA Analyzer, 3730xl DNA Analyzer
Product Line FAM, NED PET, VIC
Type Installation Standard
Label or Dye FAM, NED, PET, VIC
Quantity 1 Kit
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.