missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ GeneScan™ 600 LIZ™ dye Size Standard v2.0
Shop All Applied Biosystems Products
Click to view available options
Quantity:
800 reactions
Unit Size:
800 reactions
Description
Includes
Each kit contains enough standard for 800 analyses
The GeneScan™ 600 LIZ® Size Standard v2.0 is a fifth dye-labeled high density size standard for the reproducible sizing of fragment analysis data.
Order Info
Each
Specifications
Specifications
| Product Line | GeneScan™, LIZ™ |
| Software Type | Fragment Analysis |
| Starting Material | DNA |
| Content And Storage | Each kit contains enough standard for 800 analyses. Store in refrigerator (2–8°C). |
| For Use With (Equipment) | 310 Genetic Analyzer, 3500xL Dx Genetic Analyzer, SeqStudio Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Dx Genetic Analyzer |
| Label or Dye | LIZ |
| Product Type | Dye Size Standard V2.0 |
| Quantity | 800 reactions |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction