missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 400HD ROX™ dye Size Standard

Product Code. 15859746 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
800 Injections
9600 Injections
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15859746

Brand: Applied Biosystems™ 4310366

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Contains 21 ROX labeled single stranded DNA fragments for use as a standard

The GeneScan™ 400HD ROX™ Size Standard is a ROX™ dye-labeled size standard for the reproducible sizing of fragment analysis data. The standard contains 21 ROX™ dye-labeled DNA fragments.

The GeneScan™ 400HD ROX™ Size Standard is a ROX™ dye-labeled size standard for the reproducible sizing of fragment analysis data. The standard contains 21 ROX™ dye-labeled DNA fragments.

GeneScan™ 400 HD ROX™ Size Standard is designed for sizing DNA fragments in the 50-400 nucleotides range and provides 21 labeled fragments of: 50, 50, 90, 100, 120, 150, 160, 180, 190, 200, 220, 240, 260, 280, 290, 300, 320, 340, 360, 380 and 400 nucleotides. The sizing curve generated from these fragments make the GeneScan™ 400 HD ROX™ Size Standard ideal for a variety of fragment analysis applications such as Microsatellites, Fragment Length Polymorphisms and Relative Fluorescent Quantitation. Each of the DNA fragments is labeled with the ROX™ fluorophore.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 21 ROX labeled single stranded DNA fragments for use as a standard. Store the kit at 2° to 8°C. (Do not freeze).
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
For Use With (Application) Fragment analysis
Label or Dye ROX
No. of Reactions 9600 Injections
Product Line GeneScan, ROX
Product Type Dye
Quantity 9600 Injections
Test Time Standard

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.