missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ GeneScan™ 350 TAMRA™ dye Size Standard
Shop All Applied Biosystems Products
Click to view available options
Unit Size:
Each
Description
Includes
Contains12 TAMRA labeled single stranded DNA fragments for use as a standard
The GeneScan™ 500 XL TAMRA™ Size Standard is a TAMRA™ dye-labeled size standard for the reproducible sizing of fragment analysis data.
Order Info
Each
Specifications
Specifications
| Content And Storage | Contains12 TAMRA labeled single stranded DNA fragments for use as a standard. Store the kit at 2-8°C. (Do not freeze). |
| Format | Standard |
| For Use With (Equipment) | 310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Label or Dye | TAMRA™ |
| No. of Reactions | 800 Reactions |
| Product Line | GeneScan, TAMRA |
| Product Type | Dye |
| Shipping Condition | Wet Ice |
| Test Time | Standard |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction