missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 2500 TAMRA™ dye Size Standard

Product Code. 16355242 Shop All Applied Biosystems Products
Click to view available options
:
Contains 27 TAMRA dye labeled DNA fragments for use as standard
This item is not returnable. View return policy

Product Code. 16355242

Brand: Applied Biosystems™ 401545

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The GeneScan™ 2500 TAMRA™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence-based DNA electrophoresis systems.

The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs. GeneScan™ 2500 TAMRA™ Size provides labeled fragments of 37, 94, 109, 116, 172, 186, 222, 233, 238, 269, 286, 361, 470, 490, and 536 bases as well as larger fragments. Each of the DNA fragments is labeled with the TAMRA™ fluorophore, which results in a single peak when run under denaturing or native conditions.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 27 TAMRA dye labeled DNA fragments for use as standard. Store the kit (at 2°C to 8°C. (Do not freeze).)
For Use With (Equipment) 310 Genetic Analyzer, 3130 xl Genetic Analyzer, 3730 xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500 xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Real Time PCR (qPCR)
Label or Dye TAMRA™
No. of Reactions 400 Reactions
Product Line GeneScan, TAMRA
Product Type Dye
Shipping Condition Wet Ice
Test Time Standard
Form Wet Ice
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.