Learn More
Description
Includes
Contains 68 LIZ dye labeled single stranded DNA fragments for use as fifth dye standard
The GeneScan™ 1200 LIZ™ Size Standard is a fifth dye (LIZ™) labeled high density size standard for the reproducible sizing of fragment analysis samples. The standard contains 68 LIZ™ dye-labeled, single-stranded DNA fragments. Since the standard is labeled with the fifth dye, users can genotype a greater number of markers in a given lane, compared to the four-dye system.
GeneScan™ 1200 LIZ™ Size Standard is designed for sizing DNA fragments in the 20-1200 nucleotides range and provides 68 single-stranded labeled fragments of: 20, 30, 40, 60, 80, 100, 114, 120, 140, 160, 180, 200, 214, 220, 240, 250, 260, 280, 300, 314, 320, 340, 360, 380, 400, 414, 420, 440, 460, 480, 500, 514, 520, 540, 560, 580, 600, 614, 620, 640, 660, 680, 700, 714, 720, 740, 760, 780, 800, 820, 840, 850, 860, 880, 900, 920, 940, 960, 980, 1000, 1020, 1040, 1060, 1080, 1100, 1120, 1160, 1200 bases.
Useful for Many Applications
The sizing curve generated from fragments make the GeneScan™ 1200 LIZ™ Size Standard ideal for use for a variety of large fragment analysis applications such as BAC Fingerprinting, Microsatellites, Fragment Length Polymorphisms and others. Each of the DNA fragments is labeled with the LIZ™ fluorophore which results in a single peak when run under denaturing conditions. With the 5th dye LIZ™ your marker fragments can be labeled with the dyes FAM™, VIC™ , NED™ or PET™. Each kit contains enough standard for 800 analyses.
Order Info
Each
Specifications
Specifications
| Format | Standard |
| For Use With (Equipment) | 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer |
| GC-Rich PCR Performance | High |
| For Use With (Application) | Fragment analysis |
| Label or Dye | LIZ |
| No. of Reactions | 800 |
| Product Type | Dye |
| Quantity | 800 reactions |
| Test Time | Standard |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.