missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 1000 ROX™ dye Size Standard

Product Code. 15869706 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
400 Reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15869706 400 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869706 Supplier Applied Biosystems™ Supplier No. 401098
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

This kit contains the size standard

The GeneScan™ 1000 ROX™ Size Standard is an internal lane size standard developed for use with Applied Biosystems™ fluorescence-based DNA electrophoresis systems.

The GeneScan™ 1000 ROX™ Size Standard is an internal lane size standard developed for use with Applied Biosystems™ fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs. GeneScan™ 1000 ROX™ Size Standard is designed for sizing DNA fragments in the 47–946 bp range and provides labeled fragments of 47, 51, 55, 82, 85 93 99, 126, 136, 262, 293, 317, 439, 557, 692, 695 and 946 bases. Each of the DNA fragments is labeled with ROX™ fluorophore, which results in a single peak when run under non-denaturing conditions.

Note: Under denaturing conditions using POP-4™ polymer, fragment lengths between 100 and 539 bp can be determined (the linear range of the separation with respect to length vs. scan number).

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains the size standard.
Store the kit at 2° to 8°C (Do not freeze).
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
For Use With (Application) Fragment analysis
Label or Dye ROX
No. of Reactions 400 Reactions
Product Line GeneScan, ROX
Product Type DNA Seqeuncing Size Standard
Quantity 400 Reactions
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.