missing translation for 'onlineSavingsMsg'
Learn More

Brady™ General Purpose Vinyl Labels

Product Code. 31741205 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
48.26 x 76.20 mm
Unit Size:
100 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
31741205 48.26 x 76.20 mm 100 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31741205 Supplier Brady™ Supplier No. 177765
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible vinyl film (B-7569) with ultra-aggressive adhesive sticks permanently to rough, highly textured and difficult surfaces.

  • Permanent acrylic adhesive bonds to many common surfaces.
  • Vinyl film is suitable for indoor and outdoor use and resists moisture, chemicals, and routine handling.
  • Typical outdoor service life: up to 3 years.
  • Flexible material for use with Brady portable label printers.
  • Intended for labeling containers and packaging, including for dangerous substances. Pre-printed red diamond frames support multicolor GHS-style labels in a single print pass.
  • GHS/CLP (EU, January 2009): pictograms are shown on a white background with a red diamond border.
  • Certifications: BS 5609:1986 Section 2 and Section 3, RoHS compliant to 2011/65/EU (including amendment 2015/863).
  • Requires ribbon for printing: M7-R10000.

Specifications

Certifications/Compliance BS 5609:1986 Section 2 and section 3
Adhesive Type Permanent Acrylic
Color Red on White
Finish Gloss
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
For Use With (Application) Facility Identification, GHS and Chemical Labels, Hazard Communication Identification
For Use With (Equipment) M710, BMP71
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
Model M7-37-7569-CLP4B
No. per Pack 100 Labels/Box
Shape Rectangle
Temperature Range (English) -4°F to +167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.102 mm
Dimensions (L x W) 48.26 x 76.20 mm
Length (English) 1.90 in.
Width (English) 3 in.
Width (Metric) 76.20 mm
Width (English) Printable Area 2.25 in
Width (Metric) Printable Area 57.15 mm
Height (English) 1.90 in.
Height (Metric) 81 mm
Alternate Part Number M7-37-7569-CLP4B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.