Learn More
Thermo Scientific™ General Purpose Centrifuge Rotor Packages
Product Code. 16360778 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Model:
TX-1000 Blood Tube Package
TX-1000 Cell Culture Package
TX-1000 Microplate Package
TX-200 Blood Tube Package
TX-200 Cell Culture Package
TX-400 Blood Tube Package
TX-400 Cell Culture Package
TX-750 Blood Tube Package
TX-750 Cell Culture Package
missing translation for 'unitSize'
1 unit
Product Code. 16360778
missing translation for 'mfr': Thermo Scientific™ 75016104
Everything you need for your swing out rotor in one package
General Purpose Centrifuge Rotor packages with all the pieces you need to start spinning right away, including a rotor cross, bucket, lids and adapters
TRUSTED_SUSTAINABILITY
Specifications
Material See individual centrifuge, rotor and accessory data
Type Rotor Equipment Package
Capacity Max in TX-1000: 6 Microplates, 4 x 1000 mL BioBottles (with TX-1000 Rotors)
Net Weight See individual components of package
For Use With (Equipment) Purchase of Thermo Scientific General Purpose Centrifuge required for operation
Certifications/Compliance ISO 14971, ISO 13485, ISO 9001
Includes TX-1000 with Buckets, ClickSeal Biocontainment lids, microplate Adapters
Warranty See Manual
Biocontainment Yes
Model TX-1000 Microplate Package