missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Key Industrial Equipment™ General masking tape, LxW 50m x 25mm
Brand: Key Industrial Equipment™ 192D041N
This item is not returnable.
View return policy
Description
Highly conformable general purpose masking tape that can be removed cleanly and easily. Ideal for spray masking and many other uses from the drawing studio to the factory.- Roll length : 50m
- Width : 25mm
- Pack quantity : 36
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction