Learn More
MP Biomedicals™ Geneclean Glassmilk
Product Code. 11373099 Shop All MP Biomedicals Products
Click to view available options
Packaging Quantity:
1.25 mL
Unit Size:
1.25mL
Product Code. 11373099
Brand: MP Biomedicals™ 111001204
GeneClean Glassmilk
  • Glassmilk slurry allows flexibility of reaction size and volume.
  • GeneClean Glassmilk is a specially prepared aqueous suspension of proprietary silica matrix.
  • GeneClean Glassmilk is supplied with GeneClean Kit, which will bind to radioactive isotope-labeled ssDNA or dsDNA that is purified from agarose thus resulting in purified DNA for further use.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Kits and Assays
Description Geneclean™glassmilk™ is a specially prepared aqueous suspension of proprietary silica matrix.
Packaging Quantity 1.25 mL
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.