missing translation for 'onlineSavingsMsg'
Learn More

GeneChip™ PrimeView™ Global Gene Expression Profile Assay

Product Code. p-7181133 Shop All Applied Biosystems Products
Click to view available options
Quantity:
10 Reactions, 10 Arrays
30 Reactions, 30 Arrays
Unit Size:
10 reactions
30 reactions
This item is not returnable. View return policy

Product Code. 17163019

Brand: ABI Applied Biosystems 905406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The GeneChip PrimeView Global Gene Expression Profile Assay enables expression profiling with an emphasis on established, well-annotated content.

More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the array were selected from RefSeq version 36, UniGene database 219, and full-length human mRNAs from GenBank™.

Features include:

  • Cost-effective global confirmation of pluripotency marker expression
  • Simple-to-use online analysis tool
  • Published method with over 15,000 samples analyzed
  • For use with GeneChip Scanner 3000
  • In addition, the PluriTest™ Assay now accepts PrimeView array data from GeneChip Scanner 3000.

About the PluriTest Assay

The PluriTest Assay is a bioinformatics assay that compares the transcriptional profile of a sample to an extensive reference set of >450 cell/tissue types, including 223 hESC lines, 41 iPSC lines, somatic cells, and tissues. The online analysis tool confirms pluripotency marker expression via two separate scores: Pluripotency and Novelty. The Pluripotency Score informs the user how strongly a model-based pluripotency signature is expressed in the samples analyzed. The Novelty Score indicates the general model fit for a given sample.

TRUSTED_SUSTAINABILITY

Specifications

Array Genechip probe array; Transcriptome Profiling
Content And Storage • Arrays, purification beads (Module 2): store at 2–8°C
• Module 1, PolyA kit, hybridization kit: store at -20°C
• Fragmentation buffer (Module 2): store at room temperature

Format Array
Product Line GeneChip, PrimeView
Quantity 10 Reactions, 10 Arrays
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Species Human
For Use With (Application) Stem Cell Analysis
No. of Reactions 10 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.