Learn More
GeneChip™ PrimeView™ Global Gene Expression Profile Assay
Shop All Applied Biosystems ProductsDescription
More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the array were selected from RefSeq version 36, UniGene database 219, and full-length human mRNAs from GenBank™.
Features include:
- Cost-effective global confirmation of pluripotency marker expression
- Simple-to-use online analysis tool
- Published method with over 15,000 samples analyzed
- For use with GeneChip Scanner 3000
- In addition, the PluriTest™ Assay now accepts PrimeView array data from GeneChip Scanner 3000.
About the PluriTest Assay
The PluriTest Assay is a bioinformatics assay that compares the transcriptional profile of a sample to an extensive reference set of >450 cell/tissue types, including 223 hESC lines, 41 iPSC lines, somatic cells, and tissues. The online analysis tool confirms pluripotency marker expression via two separate scores: Pluripotency and Novelty. The Pluripotency Score informs the user how strongly a model-based pluripotency signature is expressed in the samples analyzed. The Novelty Score indicates the general model fit for a given sample.
Specifications
Specifications
| Array | 16 Array plate and trays; Transcriptome Profiling |
| Content And Storage | • Arrays, purification beads (Module 2): store at 2–8°C • Module 1, PolyA kit, hybridization kit: store at -20°C • Fragmentation buffer (Module 2): store at room temperature |
| Format | Array |
| Product Line | GeneChip, PrimeView |
| Quantity | 30 Reactions, 30 Arrays |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
| Species | Human |
| For Use With (Application) | Stem Cell Analysis |
| No. of Reactions | 30 Reactions |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.