Learn More
Thermo Scientific™ GeneBLAzer™ PPARδ-UAS-bla HEK 293T Cells
Description
PPAR delta-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Peroxisome Proliferator-Activated Receptor-delta (PPAR delta) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-PPAR delta (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. PPAR delta-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of L-165041. In addition, PPAR delta-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
Specifications
Specifications
| Alternate Agonists | Estriol, Estrone, Genistein, Diethylstibestrol, Ethynylestradiol |
| Antagonists | RU486+1 nM E2, 4-OH-tam+1 nM E2, 4-OH-tamoxifen, Tamoxifen, Tam+1 nM E2, RU486 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | PPAR delta-UAS-bla HEK 293T cells (K1244) Shipping: Dry Ice Storage: -80°C (liquid nitrogen), Liquid Nitrogen |
| Detection Method | Fluorescence, Fluorescent |
| Description | Primary Agonist: 17-β-estradiol |
| For Use With (Application) | Nuclear Receptors |
| Species | Human |
| Druggable Target | Nuclear Receptors |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.