Learn More
Thermo Scientific™ GeneBLAzer™ NPSR1-A CHO-K1 DA Assay Kit
Description
The GeneBLAzer™ NPSR1-A CHO-K1 DA (Division Arrested) cells contain the human Neuropeptide S Receptor 1-Isoform A stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1129) contain a beta-lactamase (bla) reporter gene under control of the nuclear factor of activated T-cell (NFAT) response element.
The NPSR1-A CHO-K1 DA cells and NPSR1-A-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using Neuropeptide S. In addition, NPSR1-A-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary found in the Manuals and Brochures section. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
Specifications
Specifications
| Accession Number | NP_997055.1 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Division-Arrested Cells |
| Content And Storage | Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: • GeneBLAzer™ NPSR1-A CHO-K1 DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D • GeneBLAzer™ NPSR1-A CHO-K1 Assay Protocol • LiveBLAzer™-FRET B/G Loading Protocol • Certificate of Analysis The GeneBLAzer™ NPSR1-A CHO-K1 DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Detection Method | Fluorescence |
| For Use With (Application) | GPCRs |
| Gene Symbol | NPSR1-A |
| Includes | GeneBLAzer™ NPSR1-A CHO-K1 DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70 μg, Solution D, GeneBLAzer™ NPSR1-A CHO-K1 Assay Protocol, LiveBLAzer™-FRET B/G Loading Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Product Line | GeneBLAzer |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.