missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ MR DA Assay Kit

Product Code. 10083664
Change view
Click to view available options
Quantity:
1 plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10083664 1 plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083664 Supplier Thermo Scientific™ Supplier No. K1409

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™MR DA(Division Arrested) cells and MR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Mineralcorticoid Receptor (MR) fused to the NAbinding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line.

GeneBLAzer™MR DA(Division Arrested) cells and MR-UAS-bla HEK 293T cells contain the ligand-binding domain (LBD) of the human Mineralcorticoid Receptor (MR) fused to the NAbinding domain of GAL4 stably integrated in the GeneBLAzer™UAS-bla HEK 293T cell line. GeneBLAzer™UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-MR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384- well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both MR DA cells and MRUAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of aldosterone. In addition, MR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, and substrate loading time. Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists AM 580, Fenretinide, 9-cis- retinoic acid, Ch 55, TTNPB
Antagonists LE 135
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Contains sufficient division-arrested cells and substrate to assay one 384-well plate
• MR HEK 293T DA cells (K1409A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Description GeneBLAzer™ MR DA Assay Kit, 1 Plate, ∽12,000,000 cells/vial at 12,000,000 cells/mL Quantity of Cells, 293T (HEK) Cell Line, LE 135 Antagonists, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays, Cell analysis, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, nuclear receptor biology, cell-based nuclear receptor reporter assays, target-based ADME/tox assays, cellular adme assays, nuclear receptor cellular adme assays
Gene Symbol MR
Includes MR HEK 293T DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70μg
Primary Agonist All-trans-retinoic Acid
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Sample Type 293T (HEK)
Target Entry NR3C2, MR
Type Division-Arrested Cells Assay Kit
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.