Learn More
Thermo Scientific™ GeneBLAzer™ MC5R-CRE-bla CHO-K1 Cells
Description
GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells contain the human Melanocortin Receptor 5 (MC5R) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
The GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of NPD-α MSH In addition, GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
MC5R is expressed in skin, muscle, lung, liver, spleen, and adrenal tissue. MC5R is Gs-coupled and binds all of the melanocortin peptides and ACTH. A recent study suggests that MC5R is involved in fat storage in the sebocyte cells of the skin. Another study in mice links MC5R to aggressive behavior. Mice deficient in MC5R acted less aggressively when treated with alpha-MSH.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
Specifications
| Accession Number | NM_005913.1 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells • GeneBLAzer™ MC5R-CRE-bla CHO-K1 Assay Protocol • Certificate of Analysis The GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
| Description | GeneBLAzer™ MC5R-CRE-bla CHO-K1 Cells, 2 x 106 Cells, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | FRET |
| Gene Symbol | MC5R |
| Includes | GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). Includes: GeneBLAzer™ MC5R-CRE-bla CHO-K1 cells, GeneBLAzer™ MC5R-CRE-bla CHO-K1 Assay Protocol, Certificate of Analysis |
| Population Status | Single Cell Clone Origin |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.