missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ LXR alpha DA Assay Kit

Product Code. 10073474
Change view
Click to view available options
Quantity:
1 plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10073474 1 plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10073474 Supplier Thermo Scientific™ Supplier No. K1401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ LXR alpha DA (Division Arrested) cells and LXR alpha-UAS-bla HEK 293T 293 cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-alpha(LXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line.

GeneBLAzer™ LXR alpha DA (Division Arrested) cells and LXR alpha-UAS-bla HEK 293T 293 cells contain the ligand-binding domain (LBD) of the human Liver-X receptor-alpha(LXR alpha) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line. GeneBLAzer™ UAS-bla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-LXR alpha (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase.

Division Arrested (DA) cells are available in two configurations:

  • Assay kit—includes cells and sufficient substrate to analyze 1 x 384-well plate
  • Tube of cells—sufficient to analyze 10 x 384-well plates

DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both LXR alpha DA cells and LXR alpha-UAS-bla HEK 293T 293 cells are functionally validated for Z' and EC50 concentrations of TO901317. In addition, LXR alpha-UAS-bla HEK 293T 293 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists Estriol, Estrone, Genistein, Diethylstibestrol, Ethynylestradiol
Antagonists RU486+1 nM E2, 4-OH-tam+1 nM E2, 4-OH-tamoxifen, Tamoxifen, Tam+1 nM E2, RU486
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• LXR alpha HEK 293T DA cells (K1401A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Description GeneBLAzer™ LXR alpha DA Assay Kit, 1 Plate, ∽4, 800,000 No. of Cell, Liquid, Division-Arrested Cells State, 293T (HEK) Cell Line, Fluorescent Detection Method, Nuclear Receptors Target, BLA (Beta-Lactamase) Reporter, Epithelial Cell Phenotype, Adherent Growth Properties
Detection Method Fluorescence
For Use With (Application) Cellular imaging, pharma and biopharma, target and lead identification and validation
Gene ID (Entrez) 10062
Gene Symbol LXRα
Includes LXR alpha HEK 293T DA cells (K1401A) LiveBLAzer™-FRET B/G loading kit
Primary Agonist 17-β-estradiol
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Sample Type 293T (HEK)
Target Entry LXRA, LXR alpha, NR1H3
Type Division-Arrested Cells Assay Kit
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.