missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 Cells

Product Code. 10165173
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10165173 2 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10165173 Supplier Thermo Scientific™ Supplier No. K1536

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line contains the promiscuous Gα protein, Gqo5, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line.

The GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cell line contains the promiscuous Gα protein, Gqo5, stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. NFAT-bla CHO-K1 cells (Cat. No. K1534) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element stably integrated into CHO-K1 cells. The Gqo5-NFAT-bla CHO-K1 Parental Cell line is designed for the construction of Gi/o coupled GPCR cell based assays.

TRUSTED_SUSTAINABILITY

Specifications

Cell State Dividing Cells
Content And Storage GeneBLAzer™ Gqo5-NFAT-bla CHO-K1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Detection Method Fluorescence
For Use With (Application) GPCRs
Gene Symbol Gα15
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Type Dividing Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.