Learn More
Thermo Scientific™ GeneBLAzer™ GPR54 CHO-K1 DA Assay Kit
Description
This cell line is available as dividing cells by requesting a quote for K1181 from dividingcells@invitrogen.com
GeneBLAzer™ GPR54 CHO-K1 DA (Division Arrested) cells and GPR54-NFAT-bla CHO-K1 cells contain the human KiSS1 receptor (GPR54) stably integrated into the CellSensor™ NFAT-bla CHO-K1 cell line. CellSensor™ NFAT-bla CHO-K1 cells (Cat. No. K1078) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.
DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both GPR54 CHO-K1 DA cells and GPR54-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of Metastin, (Figure 1). In addition, GPR54-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
The GPR54 receptor has been shown to be involved in tumor progression and metastasis, and in the development and functional regulation of the gonads.
Specifications
Specifications
| Accession Number | NP_115940.1 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Division-Arrested Cells |
| Content And Storage | GeneBLAzer™ GPR54 CHO-K1 DA Assay Kit Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol). • GPR54 CHO-K1 DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D, 1 mL • 2 protocols • Certificate of Analysis GeneBLAzer™ GPR54 CHO-K1 DA cells Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). • GPR54 CHO-K1 DA cells • 1 protocol • Certificate of Analysis GeneBLAzer™ GPR54-NFAT-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). • GPR54-NFAT-bla CHO-K1 cells • 1 protocol • Certificate of Analysis Storage: GeneBLAzer™ GPR54 CHO-K1 DA Cells and GPR54-NFAT-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use. |
| Description | GeneBLAzer™ GPR54 CHO-K1 DA Assay Kit, 1 Plate, ∽4, 800,000 Quantity of Cell, Liquid, Division-Arrested Cells State, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Target, BLA (Beta-Lactamase) Reporter, Epithelial Cell Phenotype, Adherent |
| Detection Method | Fluorescence |
| For Use With (Application) | Cellular imaging, pharma and biopharma, target and lead identification and validation |
| Gene ID (Entrez) | 84634 |
| Gene Symbol | GPR54 |
| Includes | Cells and sufficient substrate to analyze 1 x 384-well plate) and a tube of cells sufficient to analyze 10 x 384-well plates |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.