missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ FXR DA Assay Kit

Codice prodotto. 10135214
Cambia vista
Click to view available options
Quantity:
1 plate
Dimensione della confezione:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
10135214 1 plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10135214 Fornitore Thermo Scientific™ N. del fornitore K1397

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

GeneBLAzer™ FXR DA (Division Arrested) cells and FXR-UAS-bla HEK 293T 293 cells contain the ligand-binding domain (LBD) of the human farnesoid X receptor (FXR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line.

GeneBLAzer™ FXR DA (Division Arrested) cells and FXR-UAS-bla HEK 293T 293 cells contain the ligand-binding domain (LBD) of the human farnesoid X receptor (FXR) fused to the DNA-binding domain of GAL4 stably integrated in the GeneBLAzer™ UAS-bla HEK 293T cell line. GeneBLAzer™ UASbla HEK 293T cells stably express a beta-lactamase reporter gene under the transcriptional control of an upstream activator sequence (UAS). When an agonist binds to the LBD of the GAL4 (DBD)-FXR (LBD) fusion protein, the protein binds to the UAS, resulting in expression of beta-lactamase. Division Arrested (DA) cells are available in two configurations- an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both FXR DA cells and FXR-UAS-bla HEK 293T cells are functionally validated for Z' and EC50 concentrations of chenodeoxycholic acid (CDCA). In addition, FXR-UAS-bla HEK 293T cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.

TRUSTED_SUSTAINABILITY

Specifica

Alternate Agonists GW 0742, Retinoic Acid, GW 7647, Bezafibrate
Antagonists MK886
Assay Entry Cell-based beta lactamase reporter gene
Cell State Division-Arrested Cells
Content And Storage Contains sufficient division-arrested cells and substrate to assay 1 x 384-well plate:
• FXR HEK 293T DA cells (K1397A)
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg

Shipping: Dry Ice
Storage: -80°C (liquid nitrogen)
Description GeneBLAzer™ FXR DA Assay Kit, 1 Plate, ∽9,600,000 cells/vial at 9,600,000 Cells/mL Quantity of Cells, 293T (HEK) Cell Line, MK886 Antagonists, Fluorescent Detection Method, Nuclear Receptors Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Cellular imaging, pharma and biopharma, target and lead identification and validation, target-based adme/tox assays, Cell analysis, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, nuclear receptor biology, cell-based nuclear receptor reporter assays, target-based ADME/tox assays, cellular adme assays, nuclear receptor cellular adme assays
Gene Symbol FXR
Includes FXR HEK 293T DA cells (K1397A) LiveBLAzer™-FRET B/G Loading Kit,70μg
Primary Agonist L-165, 041
Product Line GeneBLAzer
Quantity 1 plate
Reporter Gene BLA (Beta-Lactamase)
Sample Type 293T (HEK)
Target Entry FXR, NR1H4
Type Division-Arrested Cells Assay Kit
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target Nuclear Receptors
Vedi altri risultati Mostra meno risultati

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.