Learn More
Thermo Scientific™ GeneBLAzer™ FPRL-1-Gα15 CHO-K1 DA Assay Kit
Description
This cell line is available as dividing cells by requesting a quote for K1189 from dividingcells@invitrogen.com
GeneBLAzer™ FPRL-1-Gα15 CHO-K1 DA (Division Arrested) cells and FPRL-1-Gα15-NFAT-bla CHO-K1 cells contain the human Formyl Peptide-like 1 receptor (FPRL-1) stably integrated into the GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cell line. GeneBLAzer™ Gα15-NFAT-bla CHO-K1 cells (Cat. No. K1213) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element, in addition to the promiscuous G-protein, Gα15. Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.
DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both FPRL-1-Gα15 CHO-K1 DA cells and FPRL-1-Gα15-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of WKYMV(M); (Figure 1). In addition, FPRL-1-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
The FPRL-1 receptor has been implicated in a variety of inflammation and neurological disorders, including Alzheimer's disease.
Specifications
Specifications
| Accession Number | NM_001462 |
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Division-Arrested Cells |
| Content And Storage | GeneBLAzer™ FPRL-1-Gα15 CHO-K1 DA Assay Kit Each system contains sufficient division arrested (DA) cells & substrate to assay 1 x 384-well plate. (Other materials are required separately; please refer to the protocol). • FPRL-1-Gα15 CHO-K1 DA cells • LiveBLAzer™-FRET B/G Loading Kit, 70 μg • Solution D, 1 mL • 2 protocols • Certificate of Analysis GeneBLAzer™ FPRL-1-Gα15 CHO-K1 DA cells Each system contains sufficient division arrested (DA) cells to assay 10 x 384-well plates (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). • FPRL-1-Gα15 CHO-K1 DA cells • 1 protocol • Certificate of Analysis GeneBLAzer™ FPRL-1-Gα15-NFAT-bla CHO-K1 cells (LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol). • FPRL-1-Gα15-NFAT-bla CHO-K1 cells • 1 protocol • Certificate of Analysis Storage: GeneBLAzer™ FPRL-1-Gα15 CHO-K1 DA Cells and FPRL-1-Gα15-NFAT-bla CHO-K1 Cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt or thaw for immediate use. |
| Description | GeneBLAzer™ FPRL-1-Gα±15 CHO-K1 DA Assay Kit, 1 Plate, ∽2,400,000 cells/vial Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell analysis, cellular imaging, cell-based reporter assays, cell-based and lactamase assays, drug discovery and development, target and lead identification and validation, g-protein coupled receptor biology, cell-based GPCR reporter assays |
| Gene ID (Entrez) | 3269 |
| Gene Symbol | FPRL-1 |
| Includes | GeneBLAzer™ FPRL-1-Gα±15 CHO-K1 DA Assay Kit |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.