missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ CCKBR-NFAT-bla HEK293T Cells

Product Code. 10115663
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10115663 2 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10115663 Supplier Thermo Scientific™ Supplier No. K1742

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells contain the human Cholecystokinin Receptor B (CCKBR) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line.

GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells contain the human Cholecystokinin Receptor B (CCKBR) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line. CellSensor™ NFAT-bla HEK293T cells (Cat. no. K1538) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.

The GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells are functionally validated for Z' and EC50 concentrations of CCK. In addition, GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

Cholecystokinin (CCK) is widely distributed throughout the CNS and digestive system. Studies have shown CCKBR could be involved in anxiety, learning and memory, as well as growth of gastric mucosa.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_176875.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells
• GeneBLAzer™ CCKBR-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol CCKBR
Includes GeneBLAzer™ CCKBR-NFAT-bla HEK293T cells, GeneBLAzer™ CCKBR-NFAT-bla HEK293T Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry CCKBR
Type Dividing Cells
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.