missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 Cells

Product Code. 10073493
Change view
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10073493 2 x 106 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10073493 Supplier Thermo Scientific™ Supplier No. K1471

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells contain the human Complement Component 5a Receptor 1 (ADRA1B) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line.

The GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells contain the human Complement Component 5a Receptor 1 (ADRA1B) stably integrated into the GeneBLAzer™ NFAT-bla CHO-K1 cell line. GeneBLAzer™ NFAT-bla CHO-K1 (Cat. No. K1447) contains a beta-lactamase (bla) reporter gene under control of a NFAT response element stably integrated into CHO-K1 cells.

The ADRA1B-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations using established ligands. In addition, ADRA1B-NFAT-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000679.3
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells
• GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis

The GeneBLAzer™ ADRA1B-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) GPCRs
Gene Symbol ADRA1B
Includes ADRA1B-NFAT-bla CHO-K1 cells, ADRA1B-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Population Status Pool of Stable Cells
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry ADRA1B
Type CHO-K1 Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.