missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GeneArt™ pYES1L Vector with Sapphire™ Technology

Product Code. 10048669
Click to view available options
Quantity:
10 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10048669

Brand: Invitrogen™ A13287

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A ready-to-use, 9.3 Kb linearized BAC/YAC shuttle vector, used for the assembly of DNA fragments with the GeneArt™ High-Order Genetic Assembly System

GeneArt™ pYES1L Vector with Sapphire Technology™ is a ready-to-use, 9.3 Kb linearized BAC⁄YAC shuttle vector, used for the assembly of DNA fragments with the GeneArt™ High-Order Genetic Assembly System (cat#A13285 & A13286). The vector has the capacity to clone up to 110 Kb of DNA fragments. This product only includes the linearized GeneArt™ pYES1L Vector with Sapphire Technology™.

Some of the key features and elements of the GeneArt™ pYES1L Vector with Sapphire Technology™ include:
  • TRP1 to allow selection of yeast transformants in tryptophan-deficient medium
  • ARS⁄CEN origin to allow stable maintenance of the construct in yeast
  • F’ for conjugation
  • oriT for maintenance of large constructs in E.coli
  • Spectinomycin resistance for selection in E.coli
  • 6 Convenient and validated restriction sites flanking the cloning region
For Research Use Only. Not for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY

Specifications

Antibiotic Resistance Bacterial Spectinomycin
Cloning Method Seamless Cloning
Content And Storage The product contains 3 vials of linear GeneArt pYES1L Vector with Sapphire Technology at a concentration of 50 ng/mL. Vials contain enough plasmid for 10 cloning reactions using the GeneArt High-Order Genetic Assembly System (needs to be purchased separately).

Store the product at -80°C.

The vector is guaranteed stable for 6 months when properly stored.
Sample Type DNA
For Use With (Application) Cloning
Number of Fragments Up to 10 Fragments
Product Line GeneArt
Quantity 10 Reactions
Replication Origin ARS⁄CEN, oriT
Selection Marker Eukaryotic TRP1
Shipping Condition Dry Ice
Size 110 kb total (vector plus all inserts)
Vector pYES1L Linearized
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.