missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GENEART CRISPR STRINGS DNA, U6
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Use our CRISPR guide-RNA (gRNA) in silico tool to find the optimal CRISPR sequence for your genome editing goals.
Specifications
Specifications
| Description | Crispr String U7 Promoter, Liquid, Use Our CRISPR Guide-RNA (gRNA) in Silico Tool to Find the Optimal CRISPR Sequence for your Genome Editing Goals |
| Formulation | Liquid |
| Product Type | Crispr String U7 Promoter |
| Quantity | 1 Ea. |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction