missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GeneArt™ CRISPR Nuclease Vector with CD4 Enrichment Kit (with competent cells)
Click to view available options
Quantity:
10 reactions
Unit Size:
10 reactions
Description
Includes
- CRISPR CD4 Nuclease Vector, linearized
- 10X Oligonucleotide Annealing Buffer
- DNase/RNase-free Water
- 5X Ligation Buffer
- T4 DNA Ligase
- U6 Forward Sequencing Primer
- Control Double-stranded Oligonucleotide
- OneShot™ TOP10 Chemically Competent Cells
Simple, ready-to-use, all-in-one expression vector system for expression of the functional components needed for CRISPR-Cas9 genome editing in mammalian cells with a CD4 reporter. Competent cells included for added convenience and highest cloning efficiencies.
GeneArt CRISPR Nuclease Vector with CD4 Enrichment Kit is a vector system for expression of the functional components needed for CRISPR-Cas9 genome editing in mammalian cells with a CD4 reporter. The CD4 reporter enables bead-based enrichment, an option for magnetic bead-based sorting/enrichment of Cas9 & CRISPR expressing cells using Dynabeads CD4 magnetic beads. Transfection efficiency can also be tracked using anti-CD4 fluorescent antibodies. The linearized GeneArt CRISPR nuclease vectors provide a rapid and efficient way to clone double-stranded oligonucleotides encoding a crRNA representing a desired target into an expression cassette that allows sequence-specific targeting of the Cas9 nuclease. A version with competent cells is also available (Cat. No. A21177).
The GeneArt™ CRISPR Nuclease Vector system with competent cells offers
GeneArt CRISPR Nuclease Vector with CD4 Enrichment Kit is a vector system for expression of the functional components needed for CRISPR-Cas9 genome editing in mammalian cells with a CD4 reporter. The CD4 reporter enables bead-based enrichment, an option for magnetic bead-based sorting/enrichment of Cas9 & CRISPR expressing cells using Dynabeads CD4 magnetic beads. Transfection efficiency can also be tracked using anti-CD4 fluorescent antibodies. The linearized GeneArt CRISPR nuclease vectors provide a rapid and efficient way to clone double-stranded oligonucleotides encoding a crRNA representing a desired target into an expression cassette that allows sequence-specific targeting of the Cas9 nuclease. A version with competent cells is also available (Cat. No. A21177).
The GeneArt™ CRISPR Nuclease Vector system with competent cells offers
- Easy-to-design genome engineering system
- Affordable, ready-to-use cloning vectors
- Enrichment for hard to transfect or difficult to modify cell lines
Certifications
CoA
Compliance
CoA
Recommended Storage
Store in freezer (-5 to -30°C)
Specifications
Specifications
| Cloning Method | Restriction Enzyme/MCS |
| Content And Storage | • CRISPR CD4 Nuclease Vector, linearized • 10X Oligonucleotide Annealing Buffer • DNase/RNase-free Water • 5X Ligation Buffer • T4 DNA Ligase • U6 Forward Sequencing Primer • Control, double stranded oligonucleotide Store the above at -5 to -30°C. • OneShot™ TOP10 Chemically Competent Cells Store at -80°C. |
| Detection Method | Primer-probe |
| Form | Liquid |
| Format | Kit |
| Reaction Speed | Fast |
| Technique | CRISPR-Cas9 |
| Product Line | GeneArt™ |
| Product Type | Crispr Nuclease Vector With CD4 Enrichment Kit |
| Promoter | U6, CMV |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction