Learn More
CPC™ Genderless AseptiQuik G Connector with MPC Insert
Product Code. 30968726
Change view
Click to view available options
Autoclavable:
Autoclavable
Not Autoclavable
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Autoclavable Unit Size
30968726 Autoclavable Pack of 5
30968740 Not Autoclavable Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30968726 Supplier CPC™ Supplier No. AQG17MPCHT
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The AseptiQuik G MPC adapter product is designed as an AseptiQuik G connector with an MPC insert termination, allowing you to attach directly to an MPC body. To close your open systems and enable fluid transfer outside the biological safety cabinet. From open to closed in a click.
  • Moving from open format MPC components to the closed AseptiQuik connection.
  • Enables any customer to close the system with a simple, single connection. Move outside the hood/clean bench to a general manufacturing environment.
  • Minimized space constraints of having to move into the biosafety cabinet. When equipment is fixed in place this connection can enable the rotation of the AQG connector to have an ergonomic advantage to pull the membranes in the ideal position.
  • Enables modularization of systems with MPC connectors to systems with AQG connectors on them.
  • Manage inventory SKU mismatches by quickly being able to adapt connectors of different types, when an MPC is used but needs to connect to an AseptiQuik G or vice versa.
  • Eases single-use system specifications with a one-part number for both halves of the genderless connections of the AseptiQuik. Easily operate the MPC with the use of ergonomic thumb latches.
  • Intuitive three-step connection of the AseptiQuik reduces possibilities for operator error.
Specifications
Autoclavable Autoclavable
Material Polycarbonate
Overall Length (English) 1.5 in.
Overall Length (Metric) 38.1 mm
Sterility Non-sterile
Material (Seal) Silicone
Color White
Product Type Tubing Connector