Learn More
Invitrogen™ Gemtuzumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560069
 View more versions of this product
Product Code. 30284735
  • £397.00 / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Gemtuzumab ozogamicin is a recombinant humanized IgG4 kappa antibody which is conjugated with calicheamicin derivative, a cytotoxic antitumor antibiotic isolated from fermentation of Micromonospora echinospora ssp. calichensis. Gemtuzumab ozogamicin has approximately 50% of the antibody loaded with 4-6 moles calicheamicin per mole of antibodyLabel. The antibody is specifically directed against the CD33 antigen present on leukemic myeloblasts in most patients with acute myeloid leukemia (AML). By binding to the CD33 antigen on tumors, the cytotoxic agent blocks the growth of cancerous cells and causes cell death.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gemtuzumab Humanized
Recombinant Monoclonal
Unconjugated
CDP-771; CMA-676 (ADC); gemtuzumab ozogamicin; hP67.6; hP67.6-calicheamicin (ADC); WAY-CMA-676
Protein A
RUO
Human
Antibody
IgG1 κ
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized