missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geminin Antibody (GMNN/3665) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14128JF635
This item is not returnable.
View return policy
Description
Geminin Monoclonal antibody specifically detects Geminin in Human samples. It is validated for Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/ Immunofluorescence
Specifications
| Geminin | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
| Recombinant fragment of human Geminin protein (exact sequence is proprietary) (Uniprot: O75496) | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
| 51053 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence | |
| GMNN/3665 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction