missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Geminin Antibody (2H7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04293B
This item is not returnable.
View return policy
Description
Geminin Monoclonal specifically detects Geminin in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Geminin | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GMNN | |
| Recombinant human Geminin (1-209 aa) purified from E. coli | |
| 0.1 mL | |
| Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers | |
| 51053 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2H7 | |
| Western Blot, ELISA | |
| Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human Geminin mAb, clone 2H7, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Geminin protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction