missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Gemini™ AS Automated Slide Stainer

Product Code. p-6040001 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Slide Stainer, heated
Slide Stainer, unheated
Unit Size:
1 unit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15243973 Slide Stainer, heated 1 unit
15233973 Slide Stainer, unheated 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15243973 Supplier Epredia™ Supplier No. A81500001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Improve throughput with the Gemini™ AS Automated Slide Stainer, the ultimate multitasking stainer with a footprint small enough to fit in tight spaces.

The Gemini AS Automated Slide Stainer delivers high-throughput staining capabilities to the busy histology laboratory. With its new intuitive touch-screen interface, operation is easy-to-learn and simple-to-use.

The unique dual-level design, with integrated power supply and touch-screen, ensures maximum space savings. With its small footprint, the Gemini AS Stainer can be placed near the sectioning area for optimal workflow. The intelligent look-ahead control software provides the ability to run up to 12 slide racks simultaneously with minimal reduction in speed. The “urgent” feature enables a rack to be given priority for MOHS surgery or similar STAT needs. The five heated stations provide maximum throughput without the need for a separate drying oven.

Multidimensional Design

  • Two-tier design occupies minimal bench space
  • Integrated color touch-screen with intuitive software offers quick learning and simple operation
  • Scheduler software allows multiple protocols and slide racks to be run simultaneously at optimal speed
  • User easily sees progress of slides and placement of reagents with built-in LED lighting
  • Fully editable protocols and stain sets provide flexibility in programming
  • Fume extraction, battery backup and lockable slide rack ensure safety for the slides and the operator
  • 26 or 31 reagent stations accommodate multiple protocols without changing reagents
  • One-touch on-screen view of reagent status provides simple reagent monitoring

Performance

  • Battery backup with 40 minutes of continual staining power
  • Download protocols and log files via USB connectivity
  • Load-on-demand flexibility with STAT capability
  • Units available with or without five independently heated stations
  • Six running water stations
  • Four configurable doors for convenient loading and unloading
  • Fume control easily managed with carbon filters or vent adapter kit
  • Internal flow regulators and anti-siphon devices to protect water supply
  • Lockable baskets to ensure secure slides, even upside down
  • Auto-return recalls any loaded basket to the load door
  • Passcode protection to secure customized protocols
TRUSTED_SUSTAINABILITY

Specifications

Product Type Slide Stainer, heated
Slide Capacity 20 Slides/Basket
Dimensions (L x W x H) 79 x 72 x 88 cm (31 x 28 x 35 in.)
Height (English) 35 in.
Height (Metric) 88 cm
Length (English) 31 in.
Length (Metric) 79 cm
Width (English) 28 in.
Width (Metric) 72 cm
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.