missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-36721-100ul
This item is not returnable.
View return policy
Description
GEMIN6 Polyclonal antibody specifically detects GEMIN6 in Human samples. It is validated for ELISA,Western Blot
Specifications
| GEMIN6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ23459, gem (nuclear organelle) associated protein 6, gem-associated protein 6, gemin 6, gemin-6, SIP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human GEMIN6 (NP_079051.9).,, Sequence:, MSEWMKKGPLEWQDYIYKEVRVTASEKNEYKGWVLTTDPVSANIVLVNFLEDGSMSVTGIMGHAVQTVETMNEGDHRVREKLMHLFTSGDCKAYSPEDLEERKNSLKKWLEKNHIPITEQGDAPRTLCVAGVLTIDPPYGPENCSSSNEIILSRVQDLIEGHLTASQ | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 79833 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction