missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gemin 7 Monoclonal Antibody (4H6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522597
This item is not returnable.
View return policy
Description
Peptide Sequence: ECPIAQESLE SQEQRARAAL RERYLRSLLA MVGHQVSFTL HEGVRVAAHF GATDLDVANF YVSQLQTPIG VQAEALLRCS DIISYTFKP.
The protein encoded by this gene is a component of the core SMN complex, which is required for pre-mRNA splicing in the nucleus. The encoded protein is found in the nucleoplasm, in nuclear gems (Gemini of Cajal bodies), and in the cytoplasm. Three transcript variants encoding the same protein have been found for this gene.
Specifications
| Gemin 7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9H840 | |
| Gemin7 | |
| GEMIN7 (NP_078983.1, 43 a.a. ~ 131 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 4H6 | |
| Unconjugated | |
| Gemin7 | |
| 2400008I04Rik; AI120175; AI461688; gem (nuclear organelle) associated protein 7; gem nuclear organelle associated protein 7; gem-associated protein 7; gemin 7; Gemin7; gemin-7; KIAA2024; SIP3; ZNF650 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79760 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction