missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Gemin 7 Monoclonal antibody specifically detects Gemin 7 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
Specifications
| Antigen | Gemin 7 |
| Applications | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 4H6 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_078983 |
| Gene Alias | FLJ13956, gem (nuclear organelle) associated protein 7, gem-associated protein 7, gemin 7, Gemin-7, SIP3 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?