missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gemin 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578459
This item is not returnable.
View return policy
Description
Positive Control: HeLa, HepG2 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The SMN complex plays a catalyst role in the assembly of small nuclear ribonucleoproteins (snRNPs), the building blocks of the spliceosome. Thereby, plays an important role in the splicing of cellular pre-mRNAs. Most spliceosomal snRNPs contain a common set of Sm proteins SNRPB, SNRPD1, SNRPD2, SNRPD3, SNRPE, SNRPF and SNRPG that assemble in a heptameric protein ring on the Sm site of the small nuclear RNA to form the core snRNP. In the cytosol, the Sm proteins SNRPD1, SNRPD2, SNRPE, SNRPF and SNRPG are trapped in an inactive 6S pICln-Sm complex by the chaperone CLNS1A that controls the assembly of the core snRNP. Dissociation by the SMN complex of CLNS1A from the trapped Sm proteins and their transfer to an SMN-Sm complex triggers the assembly of core snRNPs and their transport to the nucleus. GEMIN5 acts as the snRNA-binding protein of the SMN complex.
Specifications
| Gemin 5 | |
| Polyclonal | |
| Unconjugated | |
| GEMIN5 | |
| AA407055; AA407208; AI451603; BB194447; C330013N08; gem (nuclear organelle) associated protein 5; gem nuclear organelle associated protein 5; gem-associated protein 5; Gemin5; GEMIN-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 216766, 25929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.43 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8BX17, Q8TEQ6 | |
| GEMIN5 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human Gemin 5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction