missing translation for 'onlineSavingsMsg'
Learn More

Gemigliptin (tartrate), TRC

Product Code. 30402812
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402812 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402812 Supplier Toronto Research Chemicals Supplier No. TRCG24857025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gemigliptin Tartrate
CAS 1374639-74-3
Molecular Formula C18 H19 F8 N5 O2 . C4 H6 O6
Quantity 25 mg
Synonym Gemigliptin(2R,3R)-2,3-dihydroxybutanedioate
InChI Formula InChI=1S/C18H19F8N5O2.C4H6O6/c19-16(20)3-1-12(32)31(8-16)6-9(27)5-13(33)30-4-2-10-11(7-30)28-15(18(24,25)26)29-14(10)17(21,22)23;5-1(3(7)8)2(6)4(9)10/h9H,1-8,27H2;1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1
SMILES N[C@H](CN1CC(F)(F)CCC1=O)CC(=O)N2CCc3c(C2)nc(nc3C(F)(F)F)C(F)(F)F.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC Name 1-[(2S)-2-amino-4-[2,4-bis(trifluoromethyl)-6,8-dihydro-5H-pyrido[3,4-d]pyrimidin-7-yl]-4-oxo-butyl]-5,5-difluoro-piperidin-2-one;(2R,3R)-2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 639.45
Formula Weight 639.1575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.