missing translation for 'onlineSavingsMsg'
Learn More

Gemigliptin, MedChemExpress

Product Code. 16482367
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482367 50 mg 50mg
16476586 100 mg 100mg
16406248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482367 Supplier Medchem Express Supplier No. HY14892
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gemigliptin (LC15-0444 ) is a highly selective, reversible and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. Gemigliptin exhibits potent anti-glycation properties. Gemigliptin can be used for the research of advanced glycation end products (AGE)-related diabetic complications.

Gemigliptin (LC15-0444 ) is a highly selective, reversible and competitive dipeptidyl peptidase-4 (DPP-4) inhibitor, with an IC50 of 10.3 nM for human recombinant DPP-4. Gemigliptin exhibits potent anti-glycation properties. Gemigliptin can be used for the research of advanced glycation end products (AGE)-related diabetic complications.

Specifications

Chemical Name or Material Gemigliptin
CAS 911637-19-9
Color Orange
Purity Grade Notes Research
Molecular Formula C18H19F8N5O2
Quantity 50 mg
Synonym LC15-0444
Solubility Information DMSO : 50 mg/mL (102.17 mM; Need ultrasonic)
SMILES O=C1N(C[C@@H](N)CC(N2CCC3=C(C(F)(F)F)N=C(C(F)(F)F)N=C3C2)=O)CC(F)(F)CC1
Molecular Weight (g/mol) 489.36
Formula Weight 489.36
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.