missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gemifloxacin mesylate

Product Code. 16627062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16627062 250 mg 250mg
16617062 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16627062 Supplier Thermo Scientific Chemicals Supplier No. 465972500
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 210353-53-0
Molecular Formula C18 H20 F N5 O4 . C H4 O3 S

Specifications

Quantity 250 mg
Formula Weight 485.49
Physical Form Powder
Product Type Reagent
Purity ≥97.5% (HPLC)
InChI Key JIYMVSQRGZEYAX-CWUUNJJBNA-N
SMILES CS(O)(=O)=O.CO\N=C1/CN(CC1CN)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=N1)C(O)=O
IUPAC Name 7-[(4Z)-3-(aminomethyl)-4-(methoxyimino)pyrrolidin-1-yl]-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid; methanesulfonic acid
Molecular Weight (g/mol) 485.49
Chemical Name or Material Gemifloxacin mesylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.