missing translation for 'onlineSavingsMsg'
Learn More

Gemifloxacin mesylate, MedChemExpress

Product Code. 16488576
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488576 100 mg 100mg
16428658 10 mg 10mg
16404537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488576 Supplier Medchem Express Supplier No. HYB1050
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Gemifloxacin mesylate is an oral broad-spectrum quinolone antibacterial agent, used in the treatment of acute bacterial exacerbation of chronic bronchitis, and mild-to-moderate pneumonia.

Gemifloxacin mesylate is an oral broad-spectrum quinolone antibacterial agent, used in the treatment of acute bacterial exacerbation of chronic bronchitis, and mild-to-moderate pneumonia.

Specifications

Chemical Name or Material Gemifloxacin mesylate
Physical Form Solid
CAS 210353-53-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H24FN5O7S
For Use With (Application) COVID-19-immunoregulation
Synonym SB-265805S LB-20304a
Solubility Information DMSO : 100 mg/mL (205.98 mM; Need ultrasonic) ∣H2O : 50 mg/mL (102.99 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4CC(CN)/C(C4)=N/OC)=N3)C1=O)O.CS(=O)(O)=O
Molecular Weight (g/mol) 485.49
Formula Weight 485.49
Percent Purity 98.2%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.