missing translation for 'onlineSavingsMsg'
Learn More

Gemfibrozil 1-O-Beta-Glucuronide-d6, TRC

Product Code. 30240872
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.50mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240872 0.5 mg 0.50mg
30240891 1 mg 1mg
30240681 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240872 Supplier Toronto Research Chemicals Supplier No. TRCG3057620.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1703747-47-0
Molecular Formula C21H24D6O9
Molecular Weight (g/mol) 432.49
Synonym 1-[5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoate] β-D-Glucopyranuroic Acid-d6, Gemfibrozil-d6 Glucuronide;
SMILES CC1=C(OCCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C(O[C@@H]2O[C@H](C(O)=O)[C@@H](O)[C@H](O)[C@H]2O)=O)C=C(C)C=C1

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C21H30O9/c1-11-6-7-12(2)13(10-11)28-9-5-8-21(3,4)20(27)30-19-16(24)14(22)15(23)17(29-19)18(25)26/h6-7,10,14-17,19,22-24H,5,8-9H2,1-4H3,(H,25,26)/t14-,15-,16+,17-,19-/m0/s1/i3D3,4D3
Formula Weight 432.2266
Chemical Name or Material Gemfibrozil 1-O-beta-Glucuronide-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.