missing translation for 'onlineSavingsMsg'
Learn More

Gemcitabine Hydrochloride, TRC

Product Code. 30040574
Change view
Click to view available options
Quantity:
50 mg
100 mg
2.5 g
Unit Size:
100mg
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040574 2.5 g 2.5g
30010974 50 mg 50mg
30008937 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040574 Supplier Toronto Research Chemicals Supplier No. TRCG3050002.5G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122111-03-9
Molecular Formula C9 H11 F2 N3 O4 . Cl H
Molecular Weight (g/mol) 299.66
Synonym Cytidine, 2'-deoxy-2',2'-difluoro-, hydrochloride (1:1), Cytidine, 2'-deoxy-2',2'-difluoro-, monohydrochloride (9CI), Gemcitabine hydrochloride, GEMORAL, Gemcitera, Gemsar, Gemzar, LY 188011 hydrochloride
IUPAC Name 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one;hydrochloride
SMILES Cl.NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)C2(F)F

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H11F2N3O4.ClH/c10-9(11)6(16)4(3-15)18-7(9)14-2-1-5(12)13-8(14)17;/h1-2,4,6-7,15-16H,3H2,(H2,12,13,17);1H/t4-,6-,7-;/m1./s1
Formula Weight 299.0484
Percent Purity >95%
Chemical Name or Material Gemcitabine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.