missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GEM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109761
This item is not returnable.
View return policy
Description
Immunogen sequence: MTLNNVTMRQ GTVGMQPQQQ RWSIPADGRH LMVQKEPHQY SHRNRHSATP EDHCRRSWSS DSTDSVISSE SGNTYYRVVL IGEQGVGKST LANIFAGVHD SMDSDCEVLG EDTYERTLMV DGESATIILL DMWENKGENE WLHDHCMQVG.
GEM belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction.The protein encoded by this gene belongs to the RAD/GEM family of GTP-binding proteins. It is associated with the inner face of the plasma membrane and could play a role as a regulatory protein in receptor-mediated signal transduction. Alternative splicing occurs at this locus and two transcript variants encoding the same protein have been identified.
Specifications
| GEM | |
| Polyclonal | |
| Unconjugated | |
| GEM | |
| AV020497; Gem; GTP binding protein (gene overexpressed in skeletal muscle); GTP binding protein overexpressed in skeletal muscle; GTP-binding mitogen-induced T-cell protein; GTP-binding protein GEM; GTP-binding protein overexpressed in skeletal muscle; kinase-inducible Ras-like protein; KIR; MGC26294; RAS-like protein KIR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2669 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55040 | |
| GEM | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GEM (NP_005252.1). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction