missing translation for 'onlineSavingsMsg'
Learn More

Gelsolin/GSN Antibody (893205) [DyLight 680], Novus Biologicals™

Product Code. 30647056 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30647056 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30647056 Supplier Novus Biologicals Supplier No. FAB8170Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Gelsolin/GSN Monoclonal antibody specifically detects Gelsolin/GSN in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gelsolin/GSN
Applications Immunohistochemistry, Western Blot, Gene Knock-Out
Classification Monoclonal
Clone 893205
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias Actin-depolymerizing factor, ADF, AGEL, Brevin, DKFZp313L0718, gelsolin, gelsolin (amyloidosis, Finnish type)
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Gelsolin/GSN, Met1-Ala782, Accession # P06396
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cellular Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2934
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.