missing translation for 'onlineSavingsMsg'
Learn More

Gelsemine, MedChemExpress

Product Code. 16438968
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16438968

Brand: Medchem Express HYN0388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gelsemine, an alkaloid from the Chinese herb Gelsemium elegans, is effective in mitigating chronic pain. Antinociceptive effects.

Gelsemine, an alkaloid from the Chinese herb Gelsemium elegans, is effective in mitigating chronic pain. Antinociceptive effects.

Specifications

Chemical Name or Material Gelsemine
CAS 509-15-9
Color White
Purity Grade Notes Research
Molecular Formula C20H22N2O2
Quantity 10 mg
Solubility Information DMSO : ≥ 125 mg/mL (387.72 mM)
SMILES O=C1NC2=CC=CC=C2[C@]13[C@]4([H])[C@@]5([C@]6([H])C[C@H]3OC[C@@H]6[C@H]4N(C)C5)C=C
Molecular Weight (g/mol) 322.4
Formula Weight 322.4
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis