missing translation for 'onlineSavingsMsg'
Learn More

Gellan Gum

Product Code. 15465669
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465669 1 kg 1kg
15455669 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465669 Supplier Thermo Scientific Alfa Aesar Supplier No. J63423.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

An agar substitute and gelling agent | CAS: 71010-52-1 | (C24H38O20)A

Gellan Gum is an agar substitute and gelling agent, and useful in a wide variety of immobilization matricies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gellan Gum is an agar substitute and gelling agent. It is used in the food industry as an anti-settling agent, thickener or stabilizer. Also used in cosmetic industries due to its special gelling property. It is also used in pharmaceuticals and in tissue engineering. It is used as a substitute for agar in leptospiral media. Also useful in a wide variety of immobilization matricies.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71010-52-1
Molecular Formula (C24H38O20)A
MDL Number MFCD00131909
Synonym Phytagel

Specifications

Color White
Quantity 1 kg
Merck Index 14,4383
Solubility Information Soluble in water.
Physical Form Powder
Chemical Name or Material Gellan Gum

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.