missing translation for 'onlineSavingsMsg'
Learn More

Geldanamycin, MedChemExpress

Product Code. 16458967
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16458967

Brand: Medchem Express HY15230

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Geldanamycin is a Hsp90 inhibitor with antimicrobial activity against many Gram-positive and some Gram-negative bacteria. Geldanamycin has anti-influenza virus H5N1 activities.

Geldanamycin is a Hsp90 inhibitor with antimicrobial activity against many Gram-positive and some Gram-negative bacteria. Geldanamycin has anti-influenza virus H5N1 activities.

Specifications

Chemical Name or Material Geldanamycin
Physical Form Solid
CAS 30562-34-6
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C29H40N2O9
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 50 mg/mL (89.18 mM; Need ultrasonic)
SMILES COC(C(C=C1NC(/C(C)=C/C=C/[C@H](OC)[C@@H](OC(N)=O)/C(C)=C/[C@H](C)[C@H]2O)=O)=O)=C(C[C@H](C[C@@H]2OC)C)C1=O
Molecular Weight (g/mol) 560.64
Formula Weight 560.64
Percent Purity 97.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.